Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Hepes, Crystal, Biological Buffer, 7.0-8.0 pH 100 GR
SDP

Catalog No. 5026468
Encompass_Preferred

(N-2-Hydroxyethylpiperazine-N'-2-Ethanesulfonic acid)
CAS #7365-45-9 C₈H₁₈O₄N₂S F.W. 238.30
Assay: >99%

Good's buffer.
pH: 7.0-8.0

Catalog No. 50-264-68 Supplier Electron Microscopy Sciences Supplier No. 16782
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(N-2-Hydroxyethylpiperazine-N'-2-Ethanesulfonic acid)
CAS #7365-45-9 C₈H₁₈O₄N₂S F.W. 238.30
Assay: >99%

Good's buffer.
pH: 7.0-8.0

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.