Learn More
Medchemexpress LLC Hepes-d18 | 203805-87-2 | 99.89% | 50 MG
SDP
Catalog No. 5000409956
Encompass_Preferred
HEPES-d18 is the deuterium labeled version of HEPES. HEPES is a nonvolatile zwitterionic chemical buffering agent commonly used in cell culture. It is effective within a pH range of 6.8 to 8.2 and is also recognized as a potent inducer of lysosome biogenesis.
  • Used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotope incorporated into drug molecules.
  • Potential to affect pharmacokinetic and metabolic profiles of drugs through deuteration.
Catalog No. 50-004-09956 Supplier Medchemexpress LLC Supplier No. HYD0857S50MG
HEPES-d18 is the deuterium labeled version of HEPES. HEPES is a nonvolatile zwitterionic chemical buffering agent commonly used in cell culture. It is effective within a pH range of 6.8 to 8.2 and is also recognized as a potent inducer of lysosome biogenesis.
  • Used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotope incorporated into drug molecules.
  • Potential to affect pharmacokinetic and metabolic profiles of drugs through deuteration.