Learn More
Medchemexpress LLC Hepes-d18 | 203805-87-2 | 99.9% | 100 MG
SDP
Catalog No. 5000409970
Encompass_Preferred
HEPES-d18 is the deuterium labeled HEPES, a nonvolatile zwitterionic chemical buffering agent broadly applied in cell culture. It is effective at pH 6.8 to 8.2 and is also a potent inducer of lysosome biogenesis. This compound can be used as a tracer or an internal standard for quantitative analysis.
  • Effective over a broad pH range of 6.8 to 8.2.
  • Induces lysosome biogenesis.
  • Functions as a tracer in various analyses.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-09970 Supplier Medchemexpress LLC Supplier No. HYD0857S100MG
HEPES-d18 is the deuterium labeled HEPES, a nonvolatile zwitterionic chemical buffering agent broadly applied in cell culture. It is effective at pH 6.8 to 8.2 and is also a potent inducer of lysosome biogenesis. This compound can be used as a tracer or an internal standard for quantitative analysis.
  • Effective over a broad pH range of 6.8 to 8.2.
  • Induces lysosome biogenesis.
  • Functions as a tracer in various analyses.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.