Learn More
Gold Biotechnology Inc HEPES Free Acid 5 kg
SDP
Catalog No. 5000562039
Encompass_Preferred
HEPES is a buffering agent used in biochemistry and molecular biology that was selected and described by Good et al It is a zwitterionic piperazinic buffer that is useful for a pH range of 6 8 8 2 HEPES lacks the ability to form a significant complex with most metal ions and is recommended for use as a non-coordinating buffer in solutions with metal ions
Catalog No. 50-005-62039 Supplier Gold Biotechnology Inc Supplier No. H4005
HEPES is a buffering agent used in biochemistry and molecular biology that was selected and described by Good et al It is a zwitterionic piperazinic buffer that is useful for a pH range of 6 8 8 2 HEPES lacks the ability to form a significant complex with most metal ions and is recommended for use as a non-coordinating buffer in solutions with metal ions