Learn More
Research Products International Corp HEPES, Free Acid [N-(2-Hydroxyethyl) piperazine N'-(2-ethanesulfonic acid)], 50 Grams
SDP
Catalog No. 50213364
Encompass_Preferred
HEPES, Free Acid (4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid), is a  zwitterionic buffer.
In the life science field, HEPES, Free Acid, is primarily utilized as a buffering agent to maintain a stable pH in various experimental procedures. It is particularly effective in the pH range of 6.8 to 8.2, making it suitable for applications requiring slightly acidic to neutral pH conditions.  
HEPES, Free Acid, is commonly used in cell culture media, protein purification, and enzyme assays where a specific pH range needs to be maintained. Its buffering capacity and stability within the desired pH range make it valuable in these applications.
One of the significant advantages of HEPES, Free Acid, is its compatibility with biological systems and enzymes. It exhibits minimal interference with enzymatic activity and other biological processes, ensuring reliable and accurate results. 
Catalog No. 50-213-364 Supplier Research Products International Corp Supplier No. H7503050.0
HEPES, Free Acid (4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid), is a  zwitterionic buffer.
In the life science field, HEPES, Free Acid, is primarily utilized as a buffering agent to maintain a stable pH in various experimental procedures. It is particularly effective in the pH range of 6.8 to 8.2, making it suitable for applications requiring slightly acidic to neutral pH conditions.  
HEPES, Free Acid, is commonly used in cell culture media, protein purification, and enzyme assays where a specific pH range needs to be maintained. Its buffering capacity and stability within the desired pH range make it valuable in these applications.
One of the significant advantages of HEPES, Free Acid, is its compatibility with biological systems and enzymes. It exhibits minimal interference with enzymatic activity and other biological processes, ensuring reliable and accurate results. 