Learn More
MilliporeSigma™ HEPES, Free Acid, Molecular Biology Grade, Calbiochem™,
Shop All MilliporeSigma ProductsSpecifications
Specifications
| For Use With (Application) | Molecular Biology |
| Physical Form | Crystalline Powder |
| Quantity | 25 g |
| DNase | None Detected |
| Assay Percent Range | ≥99.0% |
| Protease | None Detected |
| Solubility Information | Soluble in water |
| Formula Weight | 238.3 |
| Percent Purity | ≥99.0% (T) |
| Grade | by alkalimetric assay |
| Show More |
Safety and Handling
| Recommended Storage | 15°C to 30°C (Do not freeze). |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.