Learn More
Alkali Scientific HEPES, Free Acid [N (2 Hydroxyethyl) piperazine N' (2 ethanesulfonic acid)], 100 G
SDP
Catalog No. 5000660279
Encompass_Preferred
HEPES, Free Acid (N-(2-Hydroxyethyl) Piperazine-N'-(2-Ethanesulfonic Acid)) is a biochemical reagent commonly used as a buffering agent in biological and chemical research. The free acid form of HEPES is highly effective at maintaining pH stability in biological assays, particularly in environments where pH needs to be tightly controlled. This 100g package size is ideal for smaller laboratories or experiments that require high-purity HEPES for preparing buffer solutions. HEPES is widely used in molecular biology, cell culture, and microbiology for its low interference in biological reactions, ensuring accurate and reproducible results in a variety of applications.
Catalog No. 50-006-60279 Supplier Alkali Scientific Supplier No. EPE001CP
HEPES, Free Acid (N-(2-Hydroxyethyl) Piperazine-N'-(2-Ethanesulfonic Acid)) is a biochemical reagent commonly used as a buffering agent in biological and chemical research. The free acid form of HEPES is highly effective at maintaining pH stability in biological assays, particularly in environments where pH needs to be tightly controlled. This 100g package size is ideal for smaller laboratories or experiments that require high-purity HEPES for preparing buffer solutions. HEPES is widely used in molecular biology, cell culture, and microbiology for its low interference in biological reactions, ensuring accurate and reproducible results in a variety of applications.