Learn More
Alkali Scientific HEPES, Free Acid [N (2 Hydroxyethyl) piperazine N' (2 ethanesulfonic acid)], 5 Kg
SDP
Catalog No. 5000660276
Encompass_Preferred
HEPES, Free Acid is a powerful biochemical reagent provided in a 5kg size, ideal for large-scale applications in biological and biochemical research. It helps maintain a stable pH in the range of 6.8–8.2, which is critical for a variety of assays, including cell culture, protein purification, and enzyme assays. The reagent's excellent buffering capacity ensures that pH remains stable throughout experiments, preventing fluctuations that could affect results. Its low toxicity and high stability make it particularly useful in biological systems, offering consistent performance and reproducible results for laboratories involved in cell-based studies and biochemical analysis.
Catalog No. 50-006-60276 Supplier Alkali Scientific Supplier No. EPE008CP
HEPES, Free Acid is a powerful biochemical reagent provided in a 5kg size, ideal for large-scale applications in biological and biochemical research. It helps maintain a stable pH in the range of 6.8–8.2, which is critical for a variety of assays, including cell culture, protein purification, and enzyme assays. The reagent's excellent buffering capacity ensures that pH remains stable throughout experiments, preventing fluctuations that could affect results. Its low toxicity and high stability make it particularly useful in biological systems, offering consistent performance and reproducible results for laboratories involved in cell-based studies and biochemical analysis.