Learn More
Medchemexpress LLC HEPES hemisodium salt | 103404-87-1 | MFCD00036463 | 99.0% | 249.80 | C8H18N2O4S·1/2Na | 100 MG
SDP
Catalog No. 5000282618
Encompass_Preferred
HEPES hemisodium salt is a zwitterionic buffering reagent commonly used to maintain physiological pH in biochemical assays and cell-culture media. Supplied as a solid powder, it is intended for preparation of HEPES buffer solutions and includes storage and handling guidance for laboratory use.
  • High buffering capacity at physiological pH (6.8-8.2).
  • Available as a solid powder for convenient solution preparation.
  • Reported high purity suitable for analytical and cell-culture applications.
  • Stable with sealed storage at 4°C and protected from moisture.
  • Provided with supporting documentation (datasheet and safety information).
Catalog No. 50-002-82618 Supplier Medchemexpress LLC Supplier No. HYW129509100MG
HEPES hemisodium salt is a zwitterionic buffering reagent commonly used to maintain physiological pH in biochemical assays and cell-culture media. Supplied as a solid powder, it is intended for preparation of HEPES buffer solutions and includes storage and handling guidance for laboratory use.
  • High buffering capacity at physiological pH (6.8-8.2).
  • Available as a solid powder for convenient solution preparation.
  • Reported high purity suitable for analytical and cell-culture applications.
  • Stable with sealed storage at 4°C and protected from moisture.
  • Provided with supporting documentation (datasheet and safety information).