Learn More
Medchemexpress LLC Hepes hemisodium salt | 103404-87-1 | MFCD00036463 | 99.0% | 249.80 g/mol | C8H17N2O4S · 1/2 Na | 500 MG
SDP
Catalog No. 5000283255
Encompass_Preferred
HEPES hemisodium salt is the hemisodium salt form of HEPES, a zwitterionic buffering agent commonly used to maintain physiological pH in biological and biochemical research. It delivers reliable buffer capacity near pH 6.8-8.2 and is supplied as a solid reagent for preparation of aqueous buffers for cell culture, enzymatic assays, and protein work.
  • Provides stable buffering at pH 6.8-8.2
  • Low reactivity with biological systems
  • High assay purity suitable for analytical applications
  • Easy to dissolve for reproducible buffer preparation
  • Available in convenient lab-scale pack sizes
Catalog No. 50-002-83255 Supplier Medchemexpress LLC Supplier No. HYW129509500MG
Add to Cart
Add to Cart
HEPES hemisodium salt is the hemisodium salt form of HEPES, a zwitterionic buffering agent commonly used to maintain physiological pH in biological and biochemical research. It delivers reliable buffer capacity near pH 6.8-8.2 and is supplied as a solid reagent for preparation of aqueous buffers for cell culture, enzymatic assays, and protein work.
  • Provides stable buffering at pH 6.8-8.2
  • Low reactivity with biological systems
  • High assay purity suitable for analytical applications
  • Easy to dissolve for reproducible buffer preparation
  • Available in convenient lab-scale pack sizes