Learn More
Medchemexpress LLC HEPES hemisodium salt | 103404-87-1 | MFCD00036463 | 99.0% | 249.80 g/mol | C8H18N2O4S · 1/2 Na | 10 G
SDP
Catalog No. 5000283257
Encompass_Preferred
HEPES hemisodium salt is the hemisodium salt form of the zwitterionic buffering agent HEPES, provided at 99.0% purity. It has molecular formula C8H18N2O4S · 1/2 Na and molecular weight 249.80 g/mol. The compound is used to prepare buffers effective near physiological pH for cell culture, enzymatic assays, and general biochemical applications.
  • Provides stable buffering in the pH 6.8-8.2 range.
  • High purity suitable for biochemical and cell culture applications.
  • Readily soluble for convenient buffer preparation.
  • Useful for enzymatic assays and general laboratory pH stabilization.
Catalog No. 50-002-83257 Supplier Medchemexpress LLC Supplier No. HYW12950910G
Add to Cart
Add to Cart
HEPES hemisodium salt is the hemisodium salt form of the zwitterionic buffering agent HEPES, provided at 99.0% purity. It has molecular formula C8H18N2O4S · 1/2 Na and molecular weight 249.80 g/mol. The compound is used to prepare buffers effective near physiological pH for cell culture, enzymatic assays, and general biochemical applications.
  • Provides stable buffering in the pH 6.8-8.2 range.
  • High purity suitable for biochemical and cell culture applications.
  • Readily soluble for convenient buffer preparation.
  • Useful for enzymatic assays and general laboratory pH stabilization.