Learn More
Medchemexpress LLC HEPES, NaCl, Sucrose, pH 7.2 buffer | 100 ML
SDP
Catalog No. 5000375586
Encompass_Preferred
This physiological buffer is composed of 20 mM HEPES, 150 mM NaCl, and 240 mM Sucrose, adjusted to pH 7.2. It is primarily used to resuspend human erythrocyte ghost preparations.
  • Physiological buffer
  • Composed of HEPES, NaCl, and Sucrose
  • Adjusted to pH 7.2
  • Used to resuspend human erythrocyte ghost preparations
Catalog No. 50-003-75586 Supplier Medchemexpress LLC Supplier No. HY150350100ML
Add to Cart
Add to Cart
This physiological buffer is composed of 20 mM HEPES, 150 mM NaCl, and 240 mM Sucrose, adjusted to pH 7.2. It is primarily used to resuspend human erythrocyte ghost preparations.
  • Physiological buffer
  • Composed of HEPES, NaCl, and Sucrose
  • Adjusted to pH 7.2
  • Used to resuspend human erythrocyte ghost preparations