Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ HEPES, Powder

Catalog No. MT61034RO Shop All Corning Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MT61034RO 500 g
MT61034RM 100 g
MT61034RR 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit
Catalog No. MT61034RO Supplier Corning™ Supplier No. 61034RO
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit

An alternative to bicarbonate for buffering

Cellgro incorporates many principles developed by the pharmaceutical industry including the use of WFI quality water, raw material testing, in-process monitoring, and finished product testing.

  • pH 7.2 to 7.6
  • Molecular Weight: 238.3

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HEPES
For Use With (Application) Mammalian Cell Culture
Physical Form Powder
Quantity 500 g
Packaging Plastic Bottle
Sterility Sterile
Molecular Weight (g/mol) 238.3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.