Learn More
Research Products International Corp HEPES, Sodium Salt, 250 G
SDP
Catalog No. 502230514
Encompass_Preferred
pKa (20°C): 7.45-7.65
pH Range: 6.8-8.2
Catalog No. 50-223-0514 Supplier Research Products International Corp Supplier No. H75050250.0
pKa (20°C): 7.45-7.65
pH Range: 6.8-8.2