Learn More
Medchemexpress LLC HEPES (sodium) | 75277-39-3 | MFCD00036463 | 99.9% | 260.29 | 250 MG
SDP
Catalog No. 5000347335
Encompass_Preferred
HEPES sodium is a nonvolatile zwitterionic chemical buffering agent broadly applied in cell culture. It is effective at pH 6.8 to 8.2 and is also a potent inducer of lysosome biogenesis.
  • Nonvolatile zwitterionic chemical buffering agent
  • Broadly applied in cell culture
  • Effective at pH 6.8 to 8.2
  • Potent inducer of lysosome biogenesis
  • Maintains superhydrophilicity of titanium for at least 3 months, resulting in continuous retention of bioactivity and osteoconductivity
  • Drives lysosome biogenesis, affects MiT/TFE cytoplasmic-nuclear distribution, and disrupts global cellular transcriptional profiles, leading to the activation of a MiT/TFE-dependent lysosomal-autophagic gene network in cultured RAW264.7 cells
Catalog No. 50-003-47335 Supplier Medchemexpress LLC Supplier No. 25GHY108535
HEPES sodium is a nonvolatile zwitterionic chemical buffering agent broadly applied in cell culture. It is effective at pH 6.8 to 8.2 and is also a potent inducer of lysosome biogenesis.
  • Nonvolatile zwitterionic chemical buffering agent
  • Broadly applied in cell culture
  • Effective at pH 6.8 to 8.2
  • Potent inducer of lysosome biogenesis
  • Maintains superhydrophilicity of titanium for at least 3 months, resulting in continuous retention of bioactivity and osteoconductivity
  • Drives lysosome biogenesis, affects MiT/TFE cytoplasmic-nuclear distribution, and disrupts global cellular transcriptional profiles, leading to the activation of a MiT/TFE-dependent lysosomal-autophagic gene network in cultured RAW264.7 cells