Learn More
Research Products International Corp HEPES, Sodium Salt [N-(2-Hydroxyethyl) piperazine-N'-(2-ethanesulfonic acid)], 1 Kilogram
SDP
Catalog No. 501368298
Encompass_Preferred
pKa (20°C): 7.45-7.65
pH Range: 6.8-8.2
Catalog No. 50-136-8298 Supplier Research Products International Corp Supplier No. H750501000.0
pKa (20°C): 7.45-7.65
pH Range: 6.8-8.2