Learn More
Research Products International Corp HEPES, Sodium Salt [N-(2-Hydroxyethyl) piperazine-N'-(2-ethanesulfonic acid)], 100 Grams
SDP
Catalog No. 50488709
Encompass_Preferred
pKa (20°C): 7.45-7.65
pH Range: 6.8-8.2
Catalog No. 50-488-709 Supplier Research Products International Corp Supplier No. H75050100.0
Add to Cart
Add to Cart
pKa (20°C): 7.45-7.65
pH Range: 6.8-8.2