Learn More
Research Products International Corp HEPES, Sodium Salt, 5 KG
SDP
Catalog No. 501976917
Encompass_Preferred
pKa (20°C): 7.45-7.65
pH Range: 6.8-8.2
Catalog No. 50-197-6917 Supplier Research Products International Corp Supplier No. H750505000.0
pKa (20°C): 7.45-7.65
pH Range: 6.8-8.2