Learn More
Apexbio Technology LLC HEPES Sodium salt 75277-39-3 250g
SDP
Catalog No. 5000088627
Encompass_Preferred
HEPES Sodium salt (CAS 75277-39-3) is a zwitterionic buffering agent designed to stabilize hydrogen ion concentration and maintain physiological pH (6 8 8 2) in biological systems thereby supporting cellular function and biomolecular interactions HEPES Sodium salt exerts its biological activity primarily through non-volatile amphoteric chemical buffering In in vitro and cell-based settings HEPES Sodium salt supports cell viability and experimental consistency by minimizing pH fluctuations Based on these properties HEPES Sodium salt holds research potential in cell-based assays maintenance of cultured mammalian cells controlled in vitro experiments studies of lysosomal biogenesis and investigations of cellular metabolic mechanisms
Catalog No. 50-000-88627 Supplier Apexbio Technology LLC Supplier No. B7303250000
HEPES Sodium salt (CAS 75277-39-3) is a zwitterionic buffering agent designed to stabilize hydrogen ion concentration and maintain physiological pH (6 8 8 2) in biological systems thereby supporting cellular function and biomolecular interactions HEPES Sodium salt exerts its biological activity primarily through non-volatile amphoteric chemical buffering In in vitro and cell-based settings HEPES Sodium salt supports cell viability and experimental consistency by minimizing pH fluctuations Based on these properties HEPES Sodium salt holds research potential in cell-based assays maintenance of cultured mammalian cells controlled in vitro experiments studies of lysosomal biogenesis and investigations of cellular metabolic mechanisms