Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HEPES Sodium Salt, ≥98% (dry basis), Ultrapure, Thermo Scientific™

Catalog No. AAJ16928A1 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1kg
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ16928A1 1kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ16928A1 Supplier Thermo Scientific Supplier No. J16928A1

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 75277-39-3
MDL Number MFCD00036463
Linear Formula HO(CH2)2(C4H8N2)(CH2)2SO3Na
Molecular Formula C8H17N2NaO4S
Synonym hepes sodium salt, hepes hemisodium salt, sodium 2-4-2-hydroxyethyl piperazin-1-yl ethanesulfonate, unii-z9fto91o8a, 1-piperazineethanesulfonic acid, 4-2-hydroxyethyl-, monosodium salt, z9fto91o8a, sodium 4-2-hydroxyethyl piperazin-1-ylethanesulphonate, 4-2-hydroxyethyl piperazine-1-ethanesulfonic acid sodium salt, 4-2-hydroxyethyl piperazine-1-ethanesulfonic acid hemisodium salt
Assay Dry basis
Chemical Name or Material HEPES Sodium Salt

Specifications

Quantity 1kg
InChI Key RDZTWEVXRGYCFV-UHFFFAOYSA-M
SMILES [Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1
IUPAC Name sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate
Molecular Weight (g/mol) 260.28
PubChem CID 2724248
ChEBI CHEBI:46758
Formula Weight 260.29
Percent Purity ≥98%
Packaging Plastic bottle
EINECSNumber 278-169-7
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.