Learn More
Alkali Scientific HEPES, Sodium Salt [N (2 Hydroxyethyl) piperazine N' (2 ethanesulfonic acid)], 250 G
SDP
Catalog No. 5000660241
Encompass_Preferred
HEPES, Sodium Salt is a biochemical reagent supplied in a 250g package, offering a reliable buffer for experiments requiring precise pH control. This sodium salt form of HEPES maintains a stable pH of 6.8–8.2, which is essential for a variety of biochemical and biological applications such as cell culture, protein analysis, and enzymatic assays. HEPES sodium salt is known for its excellent buffering capacity, ensuring that pH remains constant even during extended incubation periods. It is widely used in research and laboratory environments due to its low toxicity, making it a preferred choice for maintaining stable conditions during biological experiments.
Catalog No. 50-006-60241 Supplier Alkali Scientific Supplier No. EPE011CP
HEPES, Sodium Salt is a biochemical reagent supplied in a 250g package, offering a reliable buffer for experiments requiring precise pH control. This sodium salt form of HEPES maintains a stable pH of 6.8–8.2, which is essential for a variety of biochemical and biological applications such as cell culture, protein analysis, and enzymatic assays. HEPES sodium salt is known for its excellent buffering capacity, ensuring that pH remains constant even during extended incubation periods. It is widely used in research and laboratory environments due to its low toxicity, making it a preferred choice for maintaining stable conditions during biological experiments.