Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Specifications
Specifications
| Quantity | 1 kg |
| DNase | None Detected |
| Assay Percent Range | ≥99.0% |
| Protease | None Detected |
| Solubility Information | Soluble in water |
| Formula Weight | 260.28 |
| Grade | Dried basis |
| Chemical Name or Material | HEPES, Sodium Salt |
Safety and Handling
| Recommended Storage | 15°C to 30°C (Do not freeze); Store in accordance with local regulations. Store in original container, protected from direct sunlight. Keep container tightly closed and sealed until ready for use. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?