Learn More
Sigma Aldrich Fine Chemicals Biosciences HEPES sodium salt solutin500ML
Catalog No. 502902406
Encompass
HEPES is a general-purpose zwitterionic buffer which does not bind magnesium calcium manganese(II) or copper(II) ions. HEPES has been classified as Good buffer by Dr. Norman Good and his colleagues in 1966. It is zwitterionic at biological pH and is most effective in the pH range of pH 6.8 to 8.2.
Catalog No. 50-290-2406 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. H3662500ML
HEPES is a general-purpose zwitterionic buffer which does not bind magnesium calcium manganese(II) or copper(II) ions. HEPES has been classified as Good buffer by Dr. Norman Good and his colleagues in 1966. It is zwitterionic at biological pH and is most effective in the pH range of pH 6.8 to 8.2.