Learn More
Sigma Aldrich Fine Chemicals Biosciences HEPES solution 1 M pH 7.0
Catalog No. 502907906
Encompass
HEPES has been described as one of the best all-purpose buffers available for biological research. At biological pH the molecule is zwitterionic and is effective as a buffer at pH 6.8 to 8.2 (pKa 7.55). It is typically used in cell culture at concentration between 5mM to 30 mM. HEPES has been used in a wide variety of applications including tissue culture. It is commonly used to buffer cell culture media in air. HEPES finds its usage in in vitro experiments on Mg.
Catalog No. 50-290-7906 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. H088720X100ML
HEPES has been described as one of the best all-purpose buffers available for biological research. At biological pH the molecule is zwitterionic and is effective as a buffer at pH 6.8 to 8.2 (pKa 7.55). It is typically used in cell culture at concentration between 5mM to 30 mM. HEPES has been used in a wide variety of applications including tissue culture. It is commonly used to buffer cell culture media in air. HEPES finds its usage in in vitro experiments on Mg.