Learn More
Sigma Aldrich Fine Chemicals Biosciences HEPES solution, 7365-45-9, MFCD00006158, 100mL
Catalog No. 501657162
Encompass
Empirical Formula (Hill Notation) C8H18N2O4S, Molecular Weight 238.30, 1 M, pH 7.0-7.6, sterile-filtered, BioReagent, suitable for cell culture, Synonym: N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid)
Catalog No. 50-165-7162 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. H0887100ML
Empirical Formula (Hill Notation) C8H18N2O4S, Molecular Weight 238.30, 1 M, pH 7.0-7.6, sterile-filtered, BioReagent, suitable for cell culture, Synonym: N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid)