Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HEPES solution, 1M, Sigma-Aldrich

Catalog No. 01258825 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-825 100 mL
01-258-875 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-258-825 Supplier MilliporeSigma Supelco Supplier No. SRE0065100ML
Add to Cart
Add to Cart

HEPES buffer preserves a normal physiological pH environment and is commonly used as a buffer in cell culture applications. HEPES buffer provides no nutritional benefit to cells, but adds extra buffering capacity to the cell culture particularly during experiments performed outside the carbon dioxide incubator.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HEPES solution
Concentration 1 M
pH 7.2 to 7.5
Physical Form Liquid
Quantity 100 mL
Solution Type Buffer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.