Learn More
Description
HEPES buffer preserves a normal physiological pH environment and is commonly used as a buffer in cell culture applications. HEPES buffer provides no nutritional benefit to cells, but adds extra buffering capacity to the cell culture particularly during experiments performed outside the carbon dioxide incubator.
Specifications
Specifications
| Chemical Name or Material | HEPES solution |
| Concentration | 1 M |
| For Use With (Application) | Mammalian Cell Culture |
| pH | 7.2 to 7.5 |
| Physical Form | Liquid |
| Quantity | 1 L |
| Solution Type | Buffer |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.