Learn More
EZ BIOSYSTEMS INC HepG2/C3A Cell Avalanche® Transfection Reagent (human hepatocellular carcinoma cell) 0.5ml
Supplier:  EZ BIOSYSTEMS INC EZTC3A01
Encompass
HepG2/C3A Cell Avalanche® Transfection Reagent is a new class of unique chemical formulations specifically formulated and optimized for transfecting HepG2/C3A cells. The proprietary formulation of lipids and polymers ensures the highest possible transfection efficiencies and viabilities for HepG2/C3A cells.
Catalog No. NC3791226