Learn More
Cayman Chemical HepoxIlIn A3 methyl esTR 100ug
SDP
Catalog No. 502985691
Encompass_Preferred
A metabolite of arachidonic acid; induces calcium release from intracellular stores and increases calcium influx more potently than the unesterified form
Catalog No. 50-298-5691 Supplier Cayman Chemical Supplier No. 22151100 UG
A metabolite of arachidonic acid; induces calcium release from intracellular stores and increases calcium influx more potently than the unesterified form