Learn More
Medchemexpress LLC HEPPS | 16052-06-5 | 99.0% | 25 G
SDP
Catalog No. 5000414375
Encompass_Preferred
HEPPS (EPPS) is a buffering agent with a useful pH range from 7.3 to 8.7. It reduces Aβ-aggregate-induced memory deficits and rescues cognitive deficits in mice. EPPS is orally active and penetrates the blood-brain barrier.
  • Buffering agent with a useful pH range from 7.3 to 8.7.
  • Reduces Aβ-aggregate-induced memory deficits.
  • Rescues cognitive deficits in mice.
  • Orally active.
  • Penetrates the blood-brain barrier.
Catalog No. 50-004-14375 Supplier Medchemexpress LLC Supplier No. 25GHYD0873
HEPPS (EPPS) is a buffering agent with a useful pH range from 7.3 to 8.7. It reduces Aβ-aggregate-induced memory deficits and rescues cognitive deficits in mice. EPPS is orally active and penetrates the blood-brain barrier.
  • Buffering agent with a useful pH range from 7.3 to 8.7.
  • Reduces Aβ-aggregate-induced memory deficits.
  • Rescues cognitive deficits in mice.
  • Orally active.
  • Penetrates the blood-brain barrier.