Learn More
Medchemexpress LLC HEPPS | 16052-06-5 | 99.0% | 50 G
SDP
Catalog No. 5000410688
Encompass_Preferred
HEPPS (EPPS) is a buffering agent with a useful pH range from 7.3 to 8.7. It reduces Aβ-aggregate-induced memory deficits and rescues cognitive deficits in mice. EPPS is orally active and penetrates the blood-brain barrier. It binds to amyloid-β aggregates and converts them into monomers, thereby reducing the formation of amyloid-β oligomers and plaques. This agent also rescues cell damage induced by amyloid-β aggregates.
  • Binds to amyloid-β aggregates and converts them into monomers
  • Reduces formation of amyloid-β oligomers and plaques
  • Rescues cell damage induced by amyloid-β aggregates
  • Improves cognitive dysfunction in APP/PS1 transgenic mice
  • Reduces Aβ oligomer and plaque deposition in the brain
  • Reduces glial GABA release in the brain
Catalog No. 50-004-10688 Supplier Medchemexpress LLC Supplier No. HYD087350G
HEPPS (EPPS) is a buffering agent with a useful pH range from 7.3 to 8.7. It reduces Aβ-aggregate-induced memory deficits and rescues cognitive deficits in mice. EPPS is orally active and penetrates the blood-brain barrier. It binds to amyloid-β aggregates and converts them into monomers, thereby reducing the formation of amyloid-β oligomers and plaques. This agent also rescues cell damage induced by amyloid-β aggregates.
  • Binds to amyloid-β aggregates and converts them into monomers
  • Reduces formation of amyloid-β oligomers and plaques
  • Rescues cell damage induced by amyloid-β aggregates
  • Improves cognitive dysfunction in APP/PS1 transgenic mice
  • Reduces Aβ oligomer and plaque deposition in the brain
  • Reduces glial GABA release in the brain