Learn More
Alkali Scientific HEPPS [N (2 Hydroxyethyl)piperazine N' (3 propane sulfonic acid)] [EPPS], 1 Kg
SDP
Catalog No. 5000660189
Encompass_Preferred
HEPPS (N-(2-Hydroxyethyl)piperazine-N'-(3-propane sulfonic acid)) is a biochemical reagent supplied in a 1 kg bottle. This high-quality buffering agent is commonly used in laboratory and industrial applications for maintaining stable pH conditions, particularly for enzyme assays, protein studies, and cell culture applications. HEPPS is particularly useful in biological research, as it maintains pH stability under a wide range of experimental conditions. Its use in protein purification and electrophoresis helps researchers obtain precise and consistent results. This 1kg bottle is ideal for high-throughput laboratories or research settings requiring large quantities of buffering solution.
Catalog No. 50-006-60189 Supplier Alkali Scientific Supplier No. EPP002CP
Add to Cart
Add to Cart
HEPPS (N-(2-Hydroxyethyl)piperazine-N'-(3-propane sulfonic acid)) is a biochemical reagent supplied in a 1 kg bottle. This high-quality buffering agent is commonly used in laboratory and industrial applications for maintaining stable pH conditions, particularly for enzyme assays, protein studies, and cell culture applications. HEPPS is particularly useful in biological research, as it maintains pH stability under a wide range of experimental conditions. Its use in protein purification and electrophoresis helps researchers obtain precise and consistent results. This 1kg bottle is ideal for high-throughput laboratories or research settings requiring large quantities of buffering solution.