Learn More
Alkali Scientific HEPPS [N (2 Hydroxyethyl)piperazine N' (3 propane sulfonic acid)] [EPPS], 100 G
SDP
Catalog No. 5000660370
Encompass_Preferred
HEPPS (N-(2-Hydroxyethyl)piperazine-N'-(3-propane sulfonic acid)) is a biochemical reagent available in a 100g bottle. This buffering agent is commonly used in laboratory settings to maintain a stable pH in aqueous solutions, particularly in biochemistry and molecular biology experiments. HEPPS is frequently used for enzyme assays, protein purification, and cell culture applications where precise pH control is necessary. It has a stable buffering capacity and works well at physiological pH levels, making it ideal for maintaining pH balance in a variety of biological assays. This reagent is essential for accurate experimental results in numerous biochemical processes.
Catalog No. 50-006-60370 Supplier Alkali Scientific Supplier No. EPP001CP
HEPPS (N-(2-Hydroxyethyl)piperazine-N'-(3-propane sulfonic acid)) is a biochemical reagent available in a 100g bottle. This buffering agent is commonly used in laboratory settings to maintain a stable pH in aqueous solutions, particularly in biochemistry and molecular biology experiments. HEPPS is frequently used for enzyme assays, protein purification, and cell culture applications where precise pH control is necessary. It has a stable buffering capacity and works well at physiological pH levels, making it ideal for maintaining pH balance in a variety of biological assays. This reagent is essential for accurate experimental results in numerous biochemical processes.