Learn More
Medchemexpress LLC Heppso | 68399-78-0 | 99.89% | 10 MM x 1 ML
SDP
Catalog No. 5000410067
Encompass_Preferred
HEPPSO is a zwitterionic buffer with a working pH range of 7.1-8.5. It exhibits a relatively high ability to bind copper(II) and has a pKa of 7.84 at a 2.0 mM buffer concentration.
  • Zwitterionic buffer.
  • Working pH range of 7.1-8.5.
  • Relatively high ability to bind copper(II).
  • pKa of 7.84 at 2.0 mM buffer concentration.
Catalog No. 50-004-10067 Supplier Medchemexpress LLC Supplier No. HYD087410MM/1ML
HEPPSO is a zwitterionic buffer with a working pH range of 7.1-8.5. It exhibits a relatively high ability to bind copper(II) and has a pKa of 7.84 at a 2.0 mM buffer concentration.
  • Zwitterionic buffer.
  • Working pH range of 7.1-8.5.
  • Relatively high ability to bind copper(II).
  • pKa of 7.84 at 2.0 mM buffer concentration.