Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Heppso | 68399-78-0 | 99.89% | 100 G
SDP

Catalog No. 5000409846
Encompass_Preferred

HEPPSO is a zwitterionic buffer with a working pH range of 7.1-8.5. It exhibits a relatively high ability to bind copper(II) and possesses a pKa of 7.84 at a 2.0 mM buffer concentration. It is intended for research use only and is not sold to patients.

  • Working pH range of 7.1-8.5.
  • Exhibits a relatively high ability to bind copper(II).
  • Possesses a pKa of 7.84 at a 2.0 mM buffer concentration.
  • For research use only.
  • Molecular weight: 268.33.
  • Formula: C9H20N2O5S.
  • Appearance: Solid.
  • Color: White to off-white.

Catalog No. 50-004-09846 Supplier Medchemexpress LLC Supplier No. HYD0874100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HEPPSO is a zwitterionic buffer with a working pH range of 7.1-8.5. It exhibits a relatively high ability to bind copper(II) and possesses a pKa of 7.84 at a 2.0 mM buffer concentration. It is intended for research use only and is not sold to patients.

  • Working pH range of 7.1-8.5.
  • Exhibits a relatively high ability to bind copper(II).
  • Possesses a pKa of 7.84 at a 2.0 mM buffer concentration.
  • For research use only.
  • Molecular weight: 268.33.
  • Formula: C9H20N2O5S.
  • Appearance: Solid.
  • Color: White to off-white.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.