Learn More
Medchemexpress LLC HEPPSO | 68399-78-0 | 99.9% | 25 G
SDP
Catalog No. 5000414376
Encompass_Preferred
HEPPSO is a zwitterionic buffer designed for various research applications. It operates within a working pH range of 7.1-8.5 and exhibits a relatively high ability to bind copper(II). With a pKa of 7.84 at a 2.0 mM buffer concentration, it is suitable for use in laboratory settings.
  • Zwitterionic buffer
  • Working pH range of 7.1-8.5
  • High ability to bind copper(II)
  • pKa of 7.84 at 2.0 mM buffer concentration
  • For research use only
Catalog No. 50-004-14376 Supplier Medchemexpress LLC Supplier No. 25GHYD0874
HEPPSO is a zwitterionic buffer designed for various research applications. It operates within a working pH range of 7.1-8.5 and exhibits a relatively high ability to bind copper(II). With a pKa of 7.84 at a 2.0 mM buffer concentration, it is suitable for use in laboratory settings.
  • Zwitterionic buffer
  • Working pH range of 7.1-8.5
  • High ability to bind copper(II)
  • pKa of 7.84 at 2.0 mM buffer concentration
  • For research use only