Learn More
Medchemexpress LLC Heppso | 68399-78-0 | 99.9% | 5 G
SDP
Catalog No. 5000410412
Encompass_Preferred
HEPPSO is a zwitterionic buffer with a working pH range of 7.1-8.5. It exhibits a relatively high ability to bind copper(II) and has a pKa of 7.84 at a 2.0 mM buffer concentration.
  • It is a zwitterionic buffer.
  • Its working pH range is 7.1-8.5.
  • It displays relatively high ability to bind copper(II).
  • It has a pKa of 7.84 at 2.0 mM buffer concentration.
  • Available in solid form, white to off-white in color.
  • Store powder at -20°C for 3 years or 4°C for 2 years.
  • Store in solvent at -80°C for 2 years or -20°C for 1 year.
Catalog No. 50-004-10412 Supplier Medchemexpress LLC Supplier No. HYD08745G
HEPPSO is a zwitterionic buffer with a working pH range of 7.1-8.5. It exhibits a relatively high ability to bind copper(II) and has a pKa of 7.84 at a 2.0 mM buffer concentration.
  • It is a zwitterionic buffer.
  • Its working pH range is 7.1-8.5.
  • It displays relatively high ability to bind copper(II).
  • It has a pKa of 7.84 at 2.0 mM buffer concentration.
  • Available in solid form, white to off-white in color.
  • Store powder at -20°C for 3 years or 4°C for 2 years.
  • Store in solvent at -80°C for 2 years or -20°C for 1 year.