Learn More
Enzo Life Sciences Hepsin (human), (recombinant) (125U)
SDP
Supplier:  Enzo Life Sciences ALX2017501
Encompass_Preferred
Alternative name: Transmembrane protease serine 1, TMPRSS1. Concentration: 100nM. MW: ~45kDa; a-subunit: ~15kDa; b-subunit: ~30kDa. Formulation: Liquid. In 100mM TRIS, pH 9.0, containing 200mM sodium chloride and 10% glycerol. Source: Produced in Drosophila Schneider 2 (S2) cells. Recombinant human hepsin (aa 45-417) is fused at the C-terminus to a V5-His-tag. Produced by plasmid transfection expression. Long Term Storage: -80°C.
Catalog No. 50-200-7979