Learn More
AdipoGen Heptachlor exo-epoxide
SDP
Supplier:  AdipoGen CDXH0237M050
Encompass_Preferred
Chemical. CAS 1024-57-3. Formula C10H5Cl7O. Molecular Weight 389.32. Heptachlor epoxide is a metabolite of the organochlorine pesticide Heptachlor. It is an oxidation product formed by many plants and animals, including humans, after exposure to Heptachlor. Compound can be used as analytical reference material.
Catalog No. 50-256-0696