Learn More
Cayman Chemical Heptacosanoic Acid, Purity: ≥95%, CAS: 7138-40-1, 500 mg
SDP
Catalog No. 502912700
Encompass_Preferred
An odd-chain saturated fatty acid; has been used as an internal standard for the quantification of VLCFAs in human serum; has been found in wood smoke and PM2.5 emissions
Catalog No. 50-291-2700 Supplier Cayman Chemical Supplier No. 36163500 MG
An odd-chain saturated fatty acid; has been used as an internal standard for the quantification of VLCFAs in human serum; has been found in wood smoke and PM2.5 emissions