Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Heptadecafluoro-1-iodooctane

Catalog No. 26254425G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-254-425G 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 26-254-425G Supplier Sigma Aldrich Supplier No. 26254425G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Heptadecafluoro-1-iodooctane

Specifications

CAS 507-63-1
Density 2.067 g/mL (at 20°C (literature))
Boiling Point 160°C to 161°C (lit.)
Molecular Formula C8F17I
Refractive Index n20/D 1.329 (literature)
Linear Formula CF3(CF2)7 I
MDL Number MFCD00001064
Quantity 100 mg
Synonym 1-Iodoperfluorooctane; Heptadecafluorooctyl iodide; Perfluorooctyl iodide
Molecular Weight (g/mol) 545.96
Percent Purity 98%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.