Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heptadecafluorononanoic Acid 95.0+%, TCI America™
SDP

Catalog No. H08435G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08435G 5 g
H084325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. H08435G Supplier TCI America Supplier No. H08435G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 375-95-1
Molecular Formula C9HF17O2
Molecular Weight (g/mol) 464.078
MDL Number MFCD00039605
InChI Key UZUFPBIDKMEQEQ-UHFFFAOYSA-N
Synonym perfluorononanoic acid, heptadecafluorononanoic acid, perfluoro-n-nonanoic acid, nonanoic acid, heptadecafluoro, perfluorononan-1-oic acid, pfna, heptadecafluoropelargonic acid, perfluornonansaeure, heptadecafluornonansaeure, heptadekafluornonansaeure
PubChem CID 67821
ChEBI CHEBI:38397
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecafluorononanoic acid
SMILES C(=O)(C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O

Specifications

Melting Point 65°C
Color White-Yellow
Quantity 5 g
UN Number 1759
Formula Weight 464.08
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Heptadecafluorononanoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.