Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heptadecafluorooctanesulfonic acid Solution, 100 μg/mL in Methanol, MilliporeSigma™ Supelco™

Catalog No. 111019083 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9083 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-101-9083 Supplier MilliporeSigma Supelco Supplier No. 336071ML
Add to Cart
Add to Cart
This item is not returnable. View return policy

Heptadecafluorooctanesulfonic acid belongs to the class of perfluorinated alkyl acids and is emerging as persistent organic contaminants in the environment.

Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 1763-23-1
Flash Point 11°C - Closed Cup
Linear Formula CF3(CF2)7 SO3 H
MDL Number MFCD00042454
Synonym PFOS; Perfluorooctanesulfonic acid
Type Analytical Standard
Quantity 1 mL
Formula Weight 500.13
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 100 μg/mL in methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.