Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Heptadecan-9-amine | 3241-20-1 | MFCD30742905 | >96.0% | 255.48 g/mol | C17H37N | 5 G
SDP

Catalog No. 5000282881
Encompass_Preferred

Heptadecan-9-amine is a long-chain primary aliphatic amine used as a chemical building block and research reagent in organic synthesis and biochemical applications. Supplied in laboratory-scale quantities, it should be stored refrigerated and protected from light to preserve stability.

  • Primary aliphatic C17 amine.
  • Molecular formula: C17H37N.
  • Molecular weight: 255.48 g/mol.
  • CAS number: 3241-20-1.
  • Appearance: colorless to light yellow liquid.
  • Storage: 4°C, protect from light.
  • Available in laboratory pack sizes, including 5 G.

Catalog No. 50-002-82881 Supplier Medchemexpress LLC Supplier No. HYW1447035G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Heptadecan-9-amine is a long-chain primary aliphatic amine used as a chemical building block and research reagent in organic synthesis and biochemical applications. Supplied in laboratory-scale quantities, it should be stored refrigerated and protected from light to preserve stability.

  • Primary aliphatic C17 amine.
  • Molecular formula: C17H37N.
  • Molecular weight: 255.48 g/mol.
  • CAS number: 3241-20-1.
  • Appearance: colorless to light yellow liquid.
  • Storage: 4°C, protect from light.
  • Available in laboratory pack sizes, including 5 G.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.