Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Heptadecan-9-amine (9-aminoheptadecane) | 3241-20-1 | MFCD30742905 | 255.48 g/mol | C17H37N | 25 G
SDP

Catalog No. 5000282926
Encompass_Preferred

Heptadecan-9-amine (9-aminoheptadecane) is a primary C17 alkylamine used as an intermediate and reagent in organic synthesis and biochemical research. It is a colorless liquid with molecular formula C17H37N and molecular weight 255.48 g/mol.

  • Primary C17 alkylamine for use as a chemical building block.
  • Suitable for organic and medicinal chemistry applications.
  • Colorless liquid appearance, suitable for solution-phase handling.
  • CAS number 3241-20-1 for unambiguous identification.
  • Available in laboratory pack sizes, including 25 G.
  • Store at 4°C protected from light; in solvent, store at -80°C for long-term stability.

Catalog No. 50-002-82926 Supplier Medchemexpress LLC Supplier No. HYW14470325G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Heptadecan-9-amine (9-aminoheptadecane) is a primary C17 alkylamine used as an intermediate and reagent in organic synthesis and biochemical research. It is a colorless liquid with molecular formula C17H37N and molecular weight 255.48 g/mol.

  • Primary C17 alkylamine for use as a chemical building block.
  • Suitable for organic and medicinal chemistry applications.
  • Colorless liquid appearance, suitable for solution-phase handling.
  • CAS number 3241-20-1 for unambiguous identification.
  • Available in laboratory pack sizes, including 25 G.
  • Store at 4°C protected from light; in solvent, store at -80°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.