Learn More
Medchemexpress LLC Heptadecan-9-amine (9-aminoheptadecane) | 3241-20-1 | MFCD30742905 | >96.0% | 255.48 g·mol⁻¹ | C17H37N | 1 G
SDP
Catalog No. 5000283843
Encompass_Preferred
Heptadecan-9-amine (9-aminoheptadecane) is a long-chain primary amine used as a building block and linker in organic synthesis and biochemical conjugation. The hydrophobic C17 backbone is suited to modify lipophilicity, anchor molecules in lipid environments, and change surface properties for material functionalization. Supplied as a small research quantity for laboratory use.
  • Primary amine suitable for amide formation and other conjugations.
  • Long C17 chain provides lipid-anchoring properties.
  • Useful as a synthetic building block and linker in organic chemistry.
  • Supplied in a small, research-oriented package for laboratory experiments.
  • Analytical documentation (COA) available upon request for quality verification.
Catalog No. 50-002-83843 Supplier Medchemexpress LLC Supplier No. HYW1447031G
Heptadecan-9-amine (9-aminoheptadecane) is a long-chain primary amine used as a building block and linker in organic synthesis and biochemical conjugation. The hydrophobic C17 backbone is suited to modify lipophilicity, anchor molecules in lipid environments, and change surface properties for material functionalization. Supplied as a small research quantity for laboratory use.
  • Primary amine suitable for amide formation and other conjugations.
  • Long C17 chain provides lipid-anchoring properties.
  • Useful as a synthetic building block and linker in organic chemistry.
  • Supplied in a small, research-oriented package for laboratory experiments.
  • Analytical documentation (COA) available upon request for quality verification.