Learn More
Medchemexpress LLC Heptadecanoic-17,17,17-d3 acid | 202528-95-8 | 273.47 | 50 MG
SDP
Catalog No. 5000445766
Encompass_Preferred
Heptadecanoic acid-d3 is the deuterium labeled Heptadecanoic acid, an odd chain saturated fatty acid (OCS-FA) associated with various diseases including coronary heart disease, prediabetes, type 2 diabetes, and multiple sclerosis. This product is for research use only and not sold to patients.
  • Can be used as a tracer for quantitation during drug development.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Contains stable heavy isotopes for pharmacokinetic and metabolic studies.
  • Potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-45766 Supplier Medchemexpress LLC Supplier No. HYW004284S50MG
Heptadecanoic acid-d3 is the deuterium labeled Heptadecanoic acid, an odd chain saturated fatty acid (OCS-FA) associated with various diseases including coronary heart disease, prediabetes, type 2 diabetes, and multiple sclerosis. This product is for research use only and not sold to patients.
  • Can be used as a tracer for quantitation during drug development.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Contains stable heavy isotopes for pharmacokinetic and metabolic studies.
  • Potential to affect the pharmacokinetic and metabolic profiles of drugs.