Learn More
Selleck Chemical LLC Heptadecanoic acid S3336-1g
SDP
Catalog No. 503108897
Encompass_Preferred
Heptadecanoic acid (C17 0) an odd chain saturated fatty acid significantly inhibits cell proliferation and migration while promoting apoptosis in PC 9 and PC 9/GR cells Heptadecanoic acid is associated with several diseases including the incidence of coronary heart disease prediabetes and type 2 diabetes as well as multiple sclerosis
Catalog No. 50-310-8897 Supplier Selleck Chemical LLC Supplier No. S33361G
Heptadecanoic acid (C17 0) an odd chain saturated fatty acid significantly inhibits cell proliferation and migration while promoting apoptosis in PC 9 and PC 9/GR cells Heptadecanoic acid is associated with several diseases including the incidence of coronary heart disease prediabetes and type 2 diabetes as well as multiple sclerosis