Learn More
Medchemexpress LLC Heptadecanoic acid | 506-12-7 | 270.45 | 1 ML
SDP
Catalog No. 5000445874
Encompass_Preferred
Heptadecanoic acid is an odd-chain saturated fatty acid (OCS-FA) with oral activity. It can inhibit cell proliferation and induce apoptosis, exhibiting antitumor activity. It is associated with various diseases including coronary heart disease, pre-diabetes, type 2 diabetes, and multiple sclerosis. This product is for research use only and not sold to patients.
  • Inhibits cell proliferation and induces apoptosis
  • Demonstrates antitumor activity by activating the PI3K/Akt signaling pathway
  • Associated with various diseases such as coronary heart disease and type 2 diabetes
  • Increases colonic motility in animal models
Catalog No. 50-004-45874 Supplier Medchemexpress LLC Supplier No. HYW00428410MM/1ML
Heptadecanoic acid is an odd-chain saturated fatty acid (OCS-FA) with oral activity. It can inhibit cell proliferation and induce apoptosis, exhibiting antitumor activity. It is associated with various diseases including coronary heart disease, pre-diabetes, type 2 diabetes, and multiple sclerosis. This product is for research use only and not sold to patients.
  • Inhibits cell proliferation and induces apoptosis
  • Demonstrates antitumor activity by activating the PI3K/Akt signaling pathway
  • Associated with various diseases such as coronary heart disease and type 2 diabetes
  • Increases colonic motility in animal models