Learn More
Medchemexpress LLC Heptadecanoic acid | 506-12-7 | 270.45 | 10 G
SDP
Catalog No. 5000449164
Encompass_Preferred
Heptadecanoic acid is an odd-chain saturated fatty acid (OCS-FA) with oral activity. It can inhibit cell proliferation and induce apoptosis, demonstrating antitumor activity. This compound is associated with various diseases, including coronary heart disease, pre-diabetes, type 2 diabetes, and multiple sclerosis. It is also a human endogenous metabolite.
  • Acts as an odd-chain saturated fatty acid (OCS-FA)
  • Exhibits oral activity
  • Inhibits cell proliferation
  • Induces apoptosis
  • Demonstrates antitumor activity
  • Associated with various diseases like coronary heart disease, pre-diabetes, type 2 diabetes, and multiple sclerosis
  • Functions as a human endogenous metabolite
Catalog No. 50-004-49164 Supplier Medchemexpress LLC Supplier No. HYW00428410G
Heptadecanoic acid is an odd-chain saturated fatty acid (OCS-FA) with oral activity. It can inhibit cell proliferation and induce apoptosis, demonstrating antitumor activity. This compound is associated with various diseases, including coronary heart disease, pre-diabetes, type 2 diabetes, and multiple sclerosis. It is also a human endogenous metabolite.
  • Acts as an odd-chain saturated fatty acid (OCS-FA)
  • Exhibits oral activity
  • Inhibits cell proliferation
  • Induces apoptosis
  • Demonstrates antitumor activity
  • Associated with various diseases like coronary heart disease, pre-diabetes, type 2 diabetes, and multiple sclerosis
  • Functions as a human endogenous metabolite